missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylhydroxylamine Hydrochloride 98.0+%, TCI America™

$115.51 - $385.03

Chemical Identifiers

CAS 50632-53-6
Molecular Formula C3H10ClNO
Molecular Weight (g/mol) 111.569
MDL Number MFCD00012599
InChI Key BYXUIKZQGOPKFR-UHFFFAOYSA-N
Synonym n-isopropylhydroxylamine hydrochloride, n-isopropyl hydroxylamine, n-isopropylhydroxylamine hcl, 2-propanamine, n-hydroxy-, hydrochloride, ccris 4317, i-propylhydroxylamine hydrochloride, isopropylhydroxylamine hydrochloride, n-hydroxypropan-2-amine hydrochloride, n-isopropyl hydroxylamine hydrochloride, n-isopropyl-hydroxylamine hydrochloride
PubChem CID 170873
IUPAC Name N-propan-2-ylhydroxylamine;hydrochloride
SMILES CC(C)NO.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I08771G
View Documents
TCI America
I08771G
1 g N/A
Only null left
 
I08775G
View Documents
TCI America
I08775G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

50632-53-6
111.569
BYXUIKZQGOPKFR-UHFFFAOYSA-N
170873
CC(C)NO.Cl
C3H10ClNO
MFCD00012599
n-isopropylhydroxylamine hydrochloride, n-isopropyl hydroxylamine, n-isopropylhydroxylamine hcl, 2-propanamine, n-hydroxy-, hydrochloride, ccris 4317, i-propylhydroxylamine hydrochloride, isopropylhydroxylamine hydrochloride, n-hydroxypropan-2-amine hydrochloride, n-isopropyl hydroxylamine hydrochloride, n-isopropyl-hydroxylamine hydrochloride
N-propan-2-ylhydroxylamine;hydrochloride

Specifications

50632-53-6
White
MFCD00012599
n-isopropylhydroxylamine hydrochloride, n-isopropyl hydroxylamine, n-isopropylhydroxylamine hcl, 2-propanamine, n-hydroxy-, hydrochloride, ccris 4317, i-propylhydroxylamine hydrochloride, isopropylhydroxylamine hydrochloride, n-hydroxypropan-2-amine hydrochloride, n-isopropyl hydroxylamine hydrochloride, n-isopropyl-hydroxylamine hydrochloride
CC(C)NO.Cl
111.569
111.57
Crystalline Powder
72°C
C3H10ClNO
1 g
BYXUIKZQGOPKFR-UHFFFAOYSA-N
N-propan-2-ylhydroxylamine;hydrochloride
170873
≥98.0% (N,T)
N-Isopropylhydroxylamine Hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'