accessibility menu, dialog, popup

UserName

CAS RN 50632-53-6

IUPAC Name: hydrogen N-(propan-2-yl)hydroxylamine chloride
Synonyms: hydrogen N-isopropylhydroxylamine chloride
Molecular Weight (g/mol): 111.57
Molecular Formula: C3H10ClNO
InChi Key: BYXUIKZQGOPKFR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)NO
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)