missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylhydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. I08775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08775G 5 g
I08771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08775G Supplier TCI America Supplier No. I08775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50632-53-6
Molecular Formula C3H10ClNO
Molecular Weight (g/mol) 111.569
MDL Number MFCD00012599
InChI Key BYXUIKZQGOPKFR-UHFFFAOYSA-N
Synonym n-isopropylhydroxylamine hydrochloride, n-isopropyl hydroxylamine, n-isopropylhydroxylamine hcl, 2-propanamine, n-hydroxy-, hydrochloride, ccris 4317, i-propylhydroxylamine hydrochloride, isopropylhydroxylamine hydrochloride, n-hydroxypropan-2-amine hydrochloride, n-isopropyl hydroxylamine hydrochloride, n-isopropyl-hydroxylamine hydrochloride
PubChem CID 170873
IUPAC Name N-propan-2-ylhydroxylamine;hydrochloride
SMILES CC(C)NO.Cl

Specifications

Melting Point 72°C
Color White
Quantity 5 g
Formula Weight 111.57
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Isopropylhydroxylamine Hydrochloride
TSCA No
Recommended Storage Freezer