missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylbenzylamine 98.0+%, TCI America™

Catalog No. B121625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B121625ML 25 mL
B1216500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B121625ML Supplier TCI America Supplier No. B121625ML

Chemical Identifiers

CAS 102-97-6
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00008863
InChI Key LYBKPDDZTNUNNM-UHFFFAOYSA-N
Synonym n-isopropylbenzylamine, n-benzylisopropylamine, isopropylbenzylamine, n-benzyl-n-isopropylamine, benzylisopropylamine, benzenemethanamine, n-1-methylethyl, n-benzyl isopropylamine, benzylamine, n-isopropyl, n-1-methylethyl-benzenemethanamine, benzyl propan-2-yl amine
PubChem CID 66024
IUPAC Name N-benzylpropan-2-amine
SMILES CC(C)NCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 103°C
Quantity 25 mL
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Isopropylbenzylamine