missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylbenzylamine, 97%

Catalog No. aab2324336
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2324336 500 g
AAB2324322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2324336 Supplier Thermo Scientific Chemicals Supplier No. B2324336
Only null left

CAS: 102-97-6 | C10H15N | 149.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-97-6
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00008863
InChI Key LYBKPDDZTNUNNM-UHFFFAOYSA-N
Synonym n-isopropylbenzylamine, n-benzylisopropylamine, isopropylbenzylamine, n-benzyl-n-isopropylamine, benzylisopropylamine, benzenemethanamine, n-1-methylethyl, n-benzyl isopropylamine, benzylamine, n-isopropyl, n-1-methylethyl-benzenemethanamine, benzyl propan-2-yl amine
PubChem CID 66024
IUPAC Name N-benzylpropan-2-amine
SMILES CC(C)NCC1=CC=CC=C1

Specifications

Density 0.91
Boiling Point 199°C to 201°C
Flash Point 87°C (190°F)
Refractive Index 1.502
Quantity 500 g
Formula Weight 149.24
Percent Purity 97%
Chemical Name or Material N-Isopropylbenzylamine
Hazard Category H227-H302-H314
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 203-067-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only