missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Isopropylacrylamide (stabilized with MEHQ) 98.0+%, TCI America™
$100.67 - $436.19
Chemical Identifiers
| CAS | 2210-25-5 |
|---|---|
| Molecular Formula | C6H11NO |
| Molecular Weight (g/mol) | 113.16 |
| MDL Number | MFCD00041913 |
| InChI Key | QNILTEGFHQSKFF-UHFFFAOYSA-N |
| Synonym | n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u |
| PubChem CID | 16637 |
| IUPAC Name | N-(propan-2-yl)prop-2-enamide |
| SMILES | CC(C)NC(=O)C=C |
Chemical Identifiers
| 2210-25-5 | |
| 113.16 | |
| QNILTEGFHQSKFF-UHFFFAOYSA-N | |
| 16637 | |
| CC(C)NC(=O)C=C |
| C6H11NO | |
| MFCD00041913 | |
| n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u | |
| N-(propan-2-yl)prop-2-enamide |
Specifications
| 2210-25-5 | |
| White-Yellow | |
| C6H11NO | |
| 25 g | |
| QNILTEGFHQSKFF-UHFFFAOYSA-N | |
| N-(propan-2-yl)prop-2-enamide | |
| 16637 | |
| ≥98.0% (GC) | |
| N-Isopropylacrylamide (stabilized with MEHQ) |
| 66°C | |
| 110°C | |
| MFCD00041913 | |
| n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u | |
| CC(C)NC(=O)C=C | |
| 113.16 | |
| 113.16 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1015
RTECSNumber : AS3675000
TSCA : Yes
Recommended Storage : Refrigerator