missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Isopropylacrylamide (stabilized with MEHQ) 98.0+%, TCI America™
Supplier: TCI America I0401100G
Specifications
| N-Isopropylacrylamide (stabilized with MEHQ) | |
| 66°C | |
| 110°C | |
| MFCD00041913 | |
| n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u | |
| CC(C)NC(=O)C=C | |
| 113.16 | |
| 113.16 | |
| Crystalline Powder |
| 2210-25-5 | |
| White-Yellow | |
| C6H11NO | |
| 100 g | |
| QNILTEGFHQSKFF-UHFFFAOYSA-N | |
| N-(propan-2-yl)prop-2-enamide | |
| 16637 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2210-25-5 | |
| 113.16 | |
| QNILTEGFHQSKFF-UHFFFAOYSA-N | |
| 16637 | |
| CC(C)NC(=O)C=C |
| C6H11NO | |
| MFCD00041913 | |
| n-isopropylacrylamide, nipam, n-iso-propylacrylamide, 2-propenamide, n-1-methylethyl, isopropyl acrylamide, acrylamide, n-isopropyl, n-isopropyl acrylamide, n-1-methylethyl-2-propenamide, isopropylamid kyseliny akrylove, unii-b7gff17l9u | |
| N-(propan-2-yl)prop-2-enamide |
Safety and Handling
EINECSNumber : (2)-1015
RTECSNumber : AS3675000
TSCA : Yes
Recommended Storage : Refrigerator