missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Iodosuccinimide 98.0+%, TCI America™
Supplier: TCI America I00745G
Specifications
| N-Iodosuccinimide | |
| 201°C | |
| MFCD00005512 | |
| n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione | |
| IN1C(=O)CCC1=O | |
| 224.99 | |
| CHEBI:53204 | |
| ≥98.0% (T) |
| 516-12-1 | |
| C4H4INO2 | |
| 5 g | |
| LQZMLBORDGWNPD-UHFFFAOYSA-N | |
| 1-iodopyrrolidine-2,5-dione | |
| 120273 | |
| 224.99 | |
| Crystalline Powder |
Chemical Identifiers
| 516-12-1 | |
| 224.99 | |
| LQZMLBORDGWNPD-UHFFFAOYSA-N | |
| 120273 | |
| 1-iodopyrrolidine-2,5-dione |
| C4H4INO2 | |
| MFCD00005512 | |
| n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione | |
| CHEBI:53204 | |
| IN1C(=O)CCC1=O |
Safety and Handling
RTECSNumber : WN2817000
TSCA : No
Recommended Storage : Freezer