missing translation for 'onlineSavingsMsg'
Learn More

N-Iodosuccinimide 98.0+%, TCI America™

Catalog No. p-7131196
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00745G 5 g
I007425G 25 g
I0074100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I00745G Supplier TCI America Supplier No. I00745G

Chemical Identifiers

CAS 516-12-1
Molecular Formula C4H4INO2
Molecular Weight (g/mol) 224.99
MDL Number MFCD00005512
InChI Key LQZMLBORDGWNPD-UHFFFAOYSA-N
Synonym n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
PubChem CID 120273
ChEBI CHEBI:53204
IUPAC Name 1-iodopyrrolidine-2,5-dione
SMILES IN1C(=O)CCC1=O

Specifications

Melting Point 201°C
Quantity 5 g
Formula Weight 224.99
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Iodosuccinimide
RTECSNumber WN2817000
TSCA No
Recommended Storage Freezer