Learn More
N-Hydroxysuccinimide, 98+%
CAS: 6066-82-6 | C4H5NO3 | 115.088 g/mol
Supplier: Thermo Scientific Chemicals A1031222
Description
N-Hydroxysuccinimide is used for improved amidations in the carbodiimide method. It is also used to activate a carboxyl group and reacts with amine to form amide. It is involved in the preparation of N-hydroxymaleimide-styrene copolymer. Further, it finds use in analytical chemistry. As an additive, it is used in the carbodiimide method for improved amidations and peptide couplings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Hydroxysuccinimide is used for improved amidations in the carbodiimide method. It is also used to activate a carboxyl group and reacts with amine to form amide. It is involved in the preparation of N-hydroxymaleimide-styrene copolymer. Further, it finds use in analytical chemistry. As an additive, it is used in the carbodiimide method for improved amidations and peptide couplings.
Solubility
Soluble in water, dimethyl formamide, alcohols and ethyl acetate. Insoluble in cold ether.
Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and strong bases. Store in a cool place.
Specifications
| N-Hydroxysuccinimide | |
| 94°C to 99°C | |
| C4H5NO3 | |
| 100 g | |
| Hygroscopic | |
| Soluble in water,dimethyl formamide,alcohols and ethyl acetate. Insoluble in cold ether. | |
| C1CC(=O)N(C1=O)O | |
| 115.088 | |
| 115.09 |
| 6066-82-6 | |
| >98% | |
| MFCD00005516 | |
| 113913 | |
| n-hydroxysuccinimide, hosu, 1-hydroxysuccinimide, 2,5-pyrrolidinedione, 1-hydroxy, 1-hydroxy-2,5-pyrrolidinedione, succinimide, n-hydroxy, n-hydroxysuccinimde, n-hydroxysuccinimid, unii-mje3791m4t, ccris 2604 | |
| NQTADLQHYWFPDB-UHFFFAOYSA-N | |
| 1-hydroxypyrrolidine-2,5-dione | |
| 80170 | |
| ≥98% |
Chemical Identifiers
| 6066-82-6 | |
| 115.088 | |
| NQTADLQHYWFPDB-UHFFFAOYSA-N | |
| 80170 | |
| C1CC(=O)N(C1=O)O |
| C4H5NO3 | |
| MFCD00005516 | |
| n-hydroxysuccinimide, hosu, 1-hydroxysuccinimide, 2,5-pyrrolidinedione, 1-hydroxy, 1-hydroxy-2,5-pyrrolidinedione, succinimide, n-hydroxy, n-hydroxysuccinimde, n-hydroxysuccinimid, unii-mje3791m4t, ccris 2604 | |
| 1-hydroxypyrrolidine-2,5-dione |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-001-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only