missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxysuccinimide, 98+%

Catalog No. p-7080677
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1031214 25 g
AAA1031222 100 g
AAA1031236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1031214 Supplier Thermo Scientific Chemicals Supplier No. A1031214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6066-82-6 | C4H5NO3 | 115.088 g/mol

N-Hydroxysuccinimide is used for improved amidations in the carbodiimide method. It is also used to activate a carboxyl group and reacts with amine to form amide. It is involved in the preparation of N-hydroxymaleimide-styrene copolymer. Further, it finds use in analytical chemistry. As an additive, it is used in the carbodiimide method for improved amidations and peptide couplings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Hydroxysuccinimide is used for improved amidations in the carbodiimide method. It is also used to activate a carboxyl group and reacts with amine to form amide. It is involved in the preparation of N-hydroxymaleimide-styrene copolymer. Further, it finds use in analytical chemistry. As an additive, it is used in the carbodiimide method for improved amidations and peptide couplings.

Solubility
Soluble in water, dimethyl formamide, alcohols and ethyl acetate. Insoluble in cold ether.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and strong bases. Store in a cool place.

Chemical Identifiers

CAS 6066-82-6
Molecular Formula C4H5NO3
Molecular Weight (g/mol) 115.088
MDL Number MFCD00005516
InChI Key NQTADLQHYWFPDB-UHFFFAOYSA-N
Synonym n-hydroxysuccinimide, hosu, 1-hydroxysuccinimide, 2,5-pyrrolidinedione, 1-hydroxy, 1-hydroxy-2,5-pyrrolidinedione, succinimide, n-hydroxy, n-hydroxysuccinimde, n-hydroxysuccinimid, unii-mje3791m4t, ccris 2604
PubChem CID 80170
IUPAC Name 1-hydroxypyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)O

Specifications

Melting Point 94°C to 99°C
Assay Percent Range >98%
Quantity 25 g
Beilstein 113913
Sensitivity Hygroscopic
Solubility Information Soluble in water,dimethyl formamide,alcohols and ethyl acetate. Insoluble in cold ether.
Formula Weight 115.09
Percent Purity ≥98%
Chemical Name or Material N-Hydroxysuccinimide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-001-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only