missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxyphthalimide 99.0+%, TCI America™

Catalog No. p-7130523
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H039525G 25 g
H0395100G 100 g
H0395500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H039525G Supplier TCI America Supplier No. H039525G

Chemical Identifiers

CAS 524-38-9
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.13
MDL Number MFCD00005891
InChI Key CFMZSMGAMPBRBE-UHFFFAOYSA-N
Synonym n-hydroxyphthalimide, 2-hydroxyisoindoline-1,3-dione, 2-hydroxy-1h-isoindole-1,3 2h-dione, nhpi, 2-hydroxyphthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxy, phthalimide, n-hydroxy, phthaloxime, unii-bxi99m81x0, 2-hydroxyisoindolin-1,3-dione
PubChem CID 10665
IUPAC Name 2-hydroxy-2,3-dihydro-1H-isoindole-1,3-dione
SMILES ON1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 231°C
Color White-Yellow
Quantity 25 g
Formula Weight 163.13
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Hydroxyphthalimide
EINECSNumber (5)-6724
RTECSNumber TI5200000
TSCA Yes