accessibility menu, dialog, popup

UserName

HONOO

CAS RN 524-38-9

IUPAC Name: 2-hydroxy-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: N-hydroxyphthalimide
Molecular Weight (g/mol): 163.13
Molecular Formula: C8H5NO3
InChi Key: CFMZSMGAMPBRBE-UHFFFAOYSA-N
SMILES: ON1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (7)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)