missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxymethylphthalimide 98.0+%, TCI America™

Catalog No. H064825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H064825G 25 g
H0648250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H064825G Supplier TCI America Supplier No. H064825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118-29-6
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00005899
InChI Key MNSGOOCAMMSKGI-UHFFFAOYSA-N
Synonym n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
PubChem CID 8354
ChEBI CHEBI:38816
IUPAC Name 2-(hydroxymethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CO

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 25 g
Formula Weight 177.16
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Hydroxymethylphthalimide
RTECSNumber TI5270000
TSCA Yes