accessibility menu, dialog, popup

UserName

HONOO

CAS RN 118-29-6

IUPAC Name: 2-(hydroxymethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: N-(hydroxymethyl)phthalimide
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: MNSGOOCAMMSKGI-UHFFFAOYSA-N
SMILES: OCN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)