missing translation for 'onlineSavingsMsg'
Learn More

N-(Hydroxymethyl)phthalimide, 97%

Catalog No. AAB2129214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2129214 25 g
AAB2129222 100 g
AAB2129236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2129214 Supplier Thermo Scientific Chemicals Supplier No. B2129214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 118-29-6 | C9H7NO3 | 177.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-29-6
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00005899
InChI Key MNSGOOCAMMSKGI-UHFFFAOYSA-N
Synonym n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
PubChem CID 8354
ChEBI CHEBI:38816
IUPAC Name 2-(hydroxymethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CO

Specifications

Melting Point 140°C to 147°C
Quantity 25 g
Beilstein 140946
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material N-(Hydroxymethyl)phthalimide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-241-4
RTECSNumber TI5270000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only