missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Hydroxy-1,8-naphthalimide 97.0+%, TCI America™
Supplier: TCI America H10405G
Specifications
| N-Hydroxy-1,8-naphthalimide | |
| Yellow | |
| MFCD00065062 | |
| n-hydroxynaphthalimide, n-hydroxy-1,8-naphthalimide, naphthalimide, n-hydroxy, naphthalhydroxamic acid, 2-hydroxy-1h-benzo de isoquinoline-1,3 2h-dione, n-hydroxy-1,8-naphthalenedicarboximide, 2-hydroxy-1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxybenzo de isoquinoline-1,3-dione, 1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxy, unii-dzg9avw5v2 | |
| C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)O | |
| 213.192 | |
| 213.19 | |
| Crystalline Powder |
| 7797-81-1 | |
| C12H7NO3 | |
| 5 g | |
| KTWCUGUUDHJVIH-UHFFFAOYSA-N | |
| 2-hydroxybenzo[de]isoquinoline-1,3-dione | |
| 82263 | |
| ≥97.0% (HPLC,T) |
Chemical Identifiers
| 7797-81-1 | |
| 213.192 | |
| KTWCUGUUDHJVIH-UHFFFAOYSA-N | |
| 82263 | |
| C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)O |
| C12H7NO3 | |
| MFCD00065062 | |
| n-hydroxynaphthalimide, n-hydroxy-1,8-naphthalimide, naphthalimide, n-hydroxy, naphthalhydroxamic acid, 2-hydroxy-1h-benzo de isoquinoline-1,3 2h-dione, n-hydroxy-1,8-naphthalenedicarboximide, 2-hydroxy-1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxybenzo de isoquinoline-1,3-dione, 1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxy, unii-dzg9avw5v2 | |
| 2-hydroxybenzo[de]isoquinoline-1,3-dione |
Safety and Handling
TSCA : Yes