missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy-1,8-naphthalimide 97.0+%, TCI America™

Catalog No. p-7130875
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
H10405G 5 g
H104025G 25 g
2 options

Catalog No. H10405G

Supplier: TCI America H10405G

Only null left

Chemical Identifiers

CAS 7797-81-1
Molecular Formula C12H7NO3
Molecular Weight (g/mol) 213.192
MDL Number MFCD00065062
InChI Key KTWCUGUUDHJVIH-UHFFFAOYSA-N
Synonym n-hydroxynaphthalimide, n-hydroxy-1,8-naphthalimide, naphthalimide, n-hydroxy, naphthalhydroxamic acid, 2-hydroxy-1h-benzo de isoquinoline-1,3 2h-dione, n-hydroxy-1,8-naphthalenedicarboximide, 2-hydroxy-1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxybenzo de isoquinoline-1,3-dione, 1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxy, unii-dzg9avw5v2
PubChem CID 82263
IUPAC Name 2-hydroxybenzo[de]isoquinoline-1,3-dione
SMILES C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 213.19
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Hydroxy-1,8-naphthalimide