missing translation for 'onlineSavingsMsg'
Learn More

n-Hexyllithium, 2.5M (33 wt.%) solution in hexane, AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC301658000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC301658000 800 mL AcroSeal™ Glass Bottle
AC301651000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC301658000 Supplier Thermo Scientific Chemicals Supplier No. 301658000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21369-64-2 , 110-54-3
Molecular Formula C6H13Li
Molecular Weight (g/mol) 92.11
MDL Number MFCD00191446
InChI Key CETVQRFGPOGIQJ-UHFFFAOYSA-N
Synonym n-hexyllithium, lithium, hexyl, hexyllithium, hexyl lithium, hexyl-lithium, lithium hexane, n-hexyllithiurn, n-hexyllitium, n-hexyl lithium, hexli
PubChem CID 2733163
IUPAC Name lithium;hexane
SMILES [Li+].CCCCC[CH2-]

Specifications

CAS Min % 32.8, 66.8
CAS Max % 33.2, 67.2
Color Orange to Yellow
Density 0.6900g/mL
Flash Point −26°C
Packaging AcroSeal™ Glass Bottle
Linear Formula LiCH2CH2CH2CH2CH2CH3
Quantity 800 mL
Specific Gravity 0.69
Solubility Information Solubility in water: vigorous reaction.
Formula Weight 92.11
Percent Purity 32.8 to 33.2% (as HexLi)
Physical Form Solution
Chemical Name or Material n-Hexyllithium, 33 wt. % in n-hexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May cause damage to organs through p
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only