missing translation for 'onlineSavingsMsg'
Learn More

n-Hexacosane, 99%

Catalog No. p-7052061
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4246106 5 g
AA4246114 25 g
AA4246122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4246106 Supplier Thermo Scientific Chemicals Supplier No. 04246106
Only null left

CAS: 630-01-3 | C26H54 | 366.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 630-01-3
Molecular Formula C26H54
Molecular Weight (g/mol) 366.72
MDL Number MFCD00009354
InChI Key HMSWAIKSFDFLKN-UHFFFAOYSA-N
Synonym n-hexacosane, pentacosane, methyl, ch3-ch2 24-ch3, hexacosane, analytical standard, hexacosane, acmc-1b86y
PubChem CID 12407
ChEBI CHEBI:32940
IUPAC Name hexacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 56°C to 58°C
Color White
Odor Weak
Quantity 5 g
Beilstein 1705609
Solubility Information Slightly soluble in water.
Formula Weight 366.72
Percent Purity 99%
Physical Form Solid
Chemical Name or Material n-Hexacosane
EINECSNumber 211-124-1
RTECSNumber ML3705000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only