accessibility menu, dialog, popup

UserName

CAS RN 630-01-3

IUPAC Name: hexacosane
Synonyms: hexacosane
Molecular Weight (g/mol): 366.72
Molecular Formula: C26H54
InChi Key: HMSWAIKSFDFLKN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
  • (2)
  • (4)
Physical Form 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)