missing translation for 'onlineSavingsMsg'
Learn More

n-Heptylbenzene, 98%

Catalog No. AAA1296922
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1296922 100 g
AAA1296936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1296922 Supplier Thermo Scientific Chemicals Supplier No. A1296922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1078-71-3 | C13H20 | 176.303 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1078-71-3
Molecular Formula C13H20
Molecular Weight (g/mol) 176.303
MDL Number MFCD00009545
InChI Key LBNXAWYDQUGHGX-UHFFFAOYSA-N
Synonym 1-phenylheptane, benzene, heptyl, n-heptylbenzene, heptane, 1-phenyl, heptane, 1 or 7-phenyl, unii-12uc21c7sn, phenylheptane, heptyl-benzene, heptane, phenyl
PubChem CID 14115
IUPAC Name heptylbenzene
SMILES CCCCCCCC1=CC=CC=C1

Specifications

Melting Point -48°C
Density 0.86
Boiling Point 230°C to 233°C
Flash Point 95°C (203°F)
Refractive Index 1.485
Quantity 100 g
UN Number UN3082
Beilstein 1905782
Formula Weight 176.3
Percent Purity 98%
Chemical Name or Material n-Heptylbenzene
Show More Show Less
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 214-084-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only