missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Heptyl 4-hydroxybenzoate, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.
Solubility
Soluble in alcohol, phenoxyethanol, propylene glycol. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.
Solubility
Soluble in alcohol, phenoxyethanol, propylene glycol. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 46°C to 49°C |
| Quantity | 25 g |
| Solubility Information | Soluble in alcohol,phenoxyethanol,propylene glycol. Insoluble in water. |
| Formula Weight | 236.31 |
| Percent Purity | 98% |
| Chemical Name or Material | n-Heptyl 4-hydroxybenzoate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 214-115-0 |
| RTECSNumber | DH2202000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only