Learn More
n-Heptane, Spectrophotometric Grade, 99+% n-Heptane 96% min
CAS: 142-82-5 | C7H16 | 100.21 g/mol
$130.47 - $464.27
Chemical Identifiers
| CAS | 142-82-5 |
|---|---|
| Molecular Formula | C7H16 |
| Molecular Weight (g/mol) | 100.21 |
| MDL Number | MFCD00009544 |
| InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
| Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
| PubChem CID | 8900 |
| ChEBI | CHEBI:43098 |
| IUPAC Name | heptane |
| SMILES | CCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA32441K2
|
Thermo Scientific Chemicals
032441K2 |
1 L |
Each for $130.47
|
|
|||||
|
AA32441M1
|
Thermo Scientific Chemicals
032441M1 |
4 x 1 L |
Each for $464.27
|
|
|||||
|
AA32441K7
|
Thermo Scientific Chemicals
032441K7 |
4 L |
Each for $285.99
|
|
|||||
Description
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 142-82-5 | |
| 100.21 | |
| IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
| 8900 | |
| heptane |
| C7H16 | |
| MFCD00009544 | |
| n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
| CHEBI:43098 | |
| CCCCCCC |
Specifications
| °C to 91°C | |
| Colorless | |
| 0.684 | |
| 1 L | |
| Petroleum-like | |
| 1.3877 | |
| C7H16 | |
| 53 hPa (40mm Hg) at 20°C | |
| n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
| IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
| heptane | |
| 8900 | |
| 100.21 | |
| n-Heptane |
| 142-82-5 | |
| Colorless liquid | |
| 97°C to 99°C | |
| -4°C (24°F) | |
| Liquid | |
| >99% | |
| MFCD00009544 | |
| 1730763 | |
| Miscible with alcohol,chloroform and ether. Immiscible with water. | |
| CCCCCCC | |
| 100.21 | |
| CHEBI:43098 | |
| Spectrophotometric |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336-H373
DOTInformation : DOT Class: 3; Packing Group: II
EINECSNumber : 205-563-8
RTECSNumber : MI7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only