missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, Reagent Grade, Reagents

Catalog No. 01337942
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337942 500 mL
01337945 2.5 L
01337943 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337942 Supplier Labchem Inc Supplier No. C1213000500C
Only null left

n-Heptane, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym Dipropylmethane
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -90.6°C
Color Colorless
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.6795
Boiling Point 98.4°C
Grade Reagent
Chemical Name or Material n-Heptane, Reagent Grade