missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Heptane, Reagent Grade, Reagents
Specifications
Specifications
| Melting Point | -90.6°C |
| Color | Colorless |
| Quantity | 200 L |
| Packaging | Steel Drum |
| Physical Form | Liquid |
| Specific Gravity | 0.6795 |
| Boiling Point | 98.4°C |
| Grade | Reagent |
| Chemical Name or Material | n-Heptane, Reagent Grade |