missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, PRF, B&J Brand, meets requirements of ASTM D2699 and D2700, >99.75%, Honeywell Burdick & Jackson

Catalog No. 6000633
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Steel Can
Steel Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000633 19 L Steel Can
6000634 204 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000633 Supplier Honeywell Inc Supplier No. RS21019
Only null left

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

CAS Max % 100.0000%
Density 0.684 g/cm3
Quantity 19 L
Flash Point −4°C
Packaging Steel Can
UN Number UN1206
Boiling Point 98.4°C
Formula Weight 100.21
Percent Purity >99.75%
Grade Primary Reference Fuel
Chemical Name or Material n-Heptane, PRF, B&J Brand, meets requirements of ASTM D2699 and D2700, >99.75%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes