missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane HPLC, Lab Chem

Catalog No. LC149054
Change view
Click to view available options
Boiling Point:
98.4 °C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Boiling Point
LC149054 98.4 °C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC149054 Supplier LabChem Supplier No. LC149054

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • n-Heptane HPLC Solvent, Safety: Toxic, flammable, irritant

Chemical Identifiers

CAS 142-82-5
Molecular Weight (g/mol) 100.2019

Specifications

Quantity 4 L
Packaging Amber Glass
Boiling Point 98.4 °C
Percent Purity >99%
Grade HPLC
Chemical Name or Material n-Heptane HPLC