missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, HPLC grade, 99+%

Catalog No. AA22911M6
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA22911M6 4 x 4 L
AA22911AK 250 mL
AA22911K2 1 L
AA22911K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA22911M6 Supplier Thermo Scientific Chemicals Supplier No. 022911M6

CAS: 142-82-5 | C7H16 | 100.21 g/mol

n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point 91°C
Density 100.21
Quantity 4 x 4 L
Flash Point -4°C (24°F)
Physical Form Liquid
Refractive Index 1.3877
Assay Percent Range 99%
Beilstein 1730763
Boiling Point 97°C to 99°C
Solubility Information Miscible with alcohol,chloroform and ether. Immiscible with water.
Formula Weight 100.21
Grade HPLC
Chemical Name or Material n-Heptane
Show More Show Less
Hazard Category H225-H304-H315-H319-H336-H373
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 205-563-8
RTECSNumber MI7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only