missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane For spectroscopy Uvasol™, MilliporeSigma™

Catalog No. M1043660500
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1043660500 500 mL Glass Bottle
M1043662500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1043660500 Supplier Technidata Supplier No. 1.04366.0500

For spectroscopy Uvasol™

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -90.5°C
Color Colorless
Density 0.68g/cm3 at 20°C
Quantity 500 mL
Flash Point −4°C (−25°F)
Ignition Point 215°C (419°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.38
Vapor Pressure 48 hPa (20°C)
UN Number 1206
Boiling Point 97°C to 98°C
Formula Weight 100.2
Chemical Name or Material n-Heptane
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H304: May be fatal if swallowed and enters airways.
Precautionary Statement H315: Causes skin irritation.
Recommended Storage Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.