missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, EMD-Millipore

Catalog No. M1096860005
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1096860005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096860005 Supplier Technidata Supplier No. 1.09686.0005
Only null left

Hydrocarbons C7

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Packaging Glass ampoule
Explosion Limit 1 to 7 %(V)
Ignition Point 215°C
Vapor Pressure 48 hPa (20°C)
Density Density: 0.68g/cm3 (20°)
Assay Percent Range ≥99.5%
Linear Formula CH3(CH2)5CH3
Quantity 5 mL
Formula Weight 100.2
Chemical Name or Material n-Heptane
Recommended Storage Store at 15° to 25°C