missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical™

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

$1354.42 - $1354.42

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
H3604 Edge
SDS View Documents
Fisher Chemical
H3604
4 L Amber Soda Lime Glass
Each for $1,354.42
Case of 4 Each for $5,417.68
Only null left
Edge
 
Description

Description

  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 197-300nm range, to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Filtered through a sub-micron filter to remove particulates
  • Intended for research use only
Specifications

Chemical Identifiers

142-82-5
100.21
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC

Specifications

-91°C
0.005 AU max. at 270nm, 0.005 AU max. at 300nm, 0.01 AU max. at 250nm, 0.01 AU max. at 260nm, 0.05 AU max. at 230nm, 0.10 AU max. at 220nm, 0.10 AU max. at 225nm, 0.25 AU max. at 210nm, 1.00 AU max. at 197nm
0.683g/cm³
4 L
Pass Test
Liquid
1ppm max.
C7H16
MFCD00009544
0.4 mPa/s at 20°C
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CCCCCCC
100.21
CHEBI:43098
OPTIMA
142-82-5
Colorless
98°C
−4°C (25°F)
Amber Soda Lime Glass
1.3845 to 1.3865 (at 25°C)
99.0%
CH3(CH2)5CH3
48mbar at 20°C
0.01% max.
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2
n-Heptane
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
missing translation for 'documents'

missing translation for 'documents'