Learn More
n-Heptane, 99%, for biochemistry, AcroSeal™, Thermo Scientific™
Chemical Identifiers
| CAS | 142-82-5 |
|---|---|
| Molecular Formula | C7H16 |
| Molecular Weight (g/mol) | 100.21 |
| MDL Number | MFCD00009544 |
| InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
| Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
| PubChem CID | 8900 |
| ChEBI | CHEBI:43098 |
| IUPAC Name | heptane |
| SMILES | CCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC447070010
|
Thermo Scientific Chemicals
447070010 |
1L |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC447070025
|
Thermo Scientific Chemicals
447070025 |
2.5L |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 142-82-5 | |
| 100.21 | |
| IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
| 8900 | |
| heptane |
| C7H16 | |
| MFCD00009544 | |
| n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
| CHEBI:43098 | |
| CCCCCCC |
Specifications
| 142-82-5 | |
| 98.0°C | |
| 98.5% min. (GC) | |
| 1.3850 to 1.3890 | |
| MFCD00009544 | |
| 0.683 | |
| n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
| IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
| heptane | |
| 8900 | |
| CHEBI:43098 | |
| 99% | |
| n-Heptane |
| 0.6830g/mL | |
| −4°C | |
| C7H16 | |
| CH3(CH2)5CH3 | |
| 1L | |
| 15,4694 | |
| Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform | |
| CCCCCCC | |
| 100.21 | |
| 0.4 mPa.s (20°C) | |
| 100.2 | |
| Biochemistry |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap
GHS Signal Word: Danger
EINECSNumber : 205-563-8
RUO â Research Use Only