missing translation for 'onlineSavingsMsg'
Learn More

n-Heptadecane, 99%

Catalog No. p-7049937
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3155114 25 g
AA3155122 100 g
AA3155136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3155114 Supplier Thermo Scientific Chemicals Supplier No. 03155114

CAS: 629-78-7 | C17H36 | 240.475 g/mol

n-Heptadecane is used as an internal standard to study the first direct comparison of kava lactone extraction efficiencies. It plays an important role as a suspended solvent used for the extraction and concentration of essential oil. It is used as a precursor in the production of iminoctadine triacetate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-78-7
Molecular Formula C17H36
Molecular Weight (g/mol) 240.475
MDL Number MFCD00009002
InChI Key NDJKXXJCMXVBJW-UHFFFAOYSA-N
Synonym n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
PubChem CID 12398
ChEBI CHEBI:16148
IUPAC Name heptadecane
SMILES CCCCCCCCCCCCCCCCC

Specifications

Melting Point 22°C
Density 0.778
Boiling Point 302°C
Refractive Index 1.4369
Quantity 25 g
Solubility Information Immiscible with water.
Formula Weight 240.48
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material n-Heptadecane
Hazard Category H304
Hazard Statement GHS H Statement
H304;
May be fatal if swallowed and enters airways.
Precautionary Statement P301+P310-P331-P501c
EINECSNumber 211-108-4
RTECSNumber MI3550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only