missing translation for 'onlineSavingsMsg'
Learn More

N-GlcNAc-Biotin, TCI America™

Catalog No. G029750MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G029750MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G029750MG Supplier TCI America Supplier No. G029750MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1272755-69-7
Molecular Formula C19H31N3O7S
Molecular Weight (g/mol) 445.531
InChI Key FSUXTBPGXKXZDJ-NWGRMCGKSA-N
Synonym 2-Acetamido-N-biotinyl-2-deoxy-beta-D-glucopyranosylamine, 2-Acetamido-2-deoxy-beta-D-glucopyranosylaminobiotin
PubChem CID 44629936
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[(2R,3S,4R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(2-oxopropyl)oxan-2-yl]pentanamide
SMILES CC(=O)CC1C(C(C(OC1NC(=O)CCCCC2C3C(CS2)NC(=O)N3)CO)O)O

Specifications

Color White
Quantity 50 mg
Formula Weight 446.52
Physical Form Crystalline Powder
Chemical Name or Material N-GlcNAc-Biotin
TSCA No
Recommended Storage Freezer