accessibility menu, dialog, popup

UserName

CAS RN 1272755-69-7

IUPAC Name: N-[4,5-dihydroxy-6-(hydroxymethyl)-3-(2-oxopropyl)oxan-2-yl]-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide
Synonyms: N-[4,5-dihydroxy-6-(hydroxymethyl)-3-(2-oxopropyl)oxan-2-yl]-5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamide
Molecular Weight (g/mol): 445.53
Molecular Formula: C19H31N3O7S
InChi Key: FSUXTBPGXKXZDJ-UHFFFAOYNA-N
SMILES: CC(=O)CC1C(O)C(O)C(CO)OC1NC(=O)CCCCC1SCC2NC(=O)NC12
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results