missing translation for 'onlineSavingsMsg'
Learn More

N-Formylglycine Ethyl Ester 98.0+%, TCI America™

Catalog No. p-7129676
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04215G 5 g
F042125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04215G Supplier TCI America Supplier No. F04215G

Chemical Identifiers

CAS 3154-51-6
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00014125
InChI Key GMBCCEOJUWMBPF-UHFFFAOYSA-N
Synonym n-formylglycine ethyl ester, ethyl n-formylglycinate, glycine, n-formyl-, ethyl ester, ethyl formylaminoacetate, ethylformylglycine, n-formylglycineethylester, ethyl formamidoacetate, 2-formylaminoacetic acid ethyl ester, for-gly-oet, ethyl formylglycinate
PubChem CID 76615
IUPAC Name ethyl 2-formamidoacetate
SMILES CCOC(=O)CNC=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 131.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Formylglycine Ethyl Ester