accessibility menu, dialog, popup

UserName

H3COOHNO

CAS RN 3154-51-6

IUPAC Name: ethyl 2-formamidoacetate
Synonyms: ethyl 2-formamidoacetate
Molecular Weight (g/mol): 131.13
Molecular Formula: C5H9NO3
InChi Key: GMBCCEOJUWMBPF-UHFFFAOYSA-N
SMILES: CCOC(=O)CNC=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results