missing translation for 'onlineSavingsMsg'
Learn More

N-Formylglycine, 98%

Catalog No. AAH3105303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3105303 1 g
AAH3105306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3105303 Supplier Thermo Scientific Chemicals Supplier No. H3105303

CAS: 2491-15-8 | C3H5NO3 | 103.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2491-15-8
Molecular Formula C3H5NO3
Molecular Weight (g/mol) 103.08
MDL Number MFCD00037360
InChI Key UGJBHEZMOKVTIM-UHFFFAOYSA-N
Synonym n-formylglycine, formylglycine, glycine, n-formyl, fgly, for-gly-oh, formylglycin, n-hydroxymethyleneglycine, formylamino acetic acid, n-formylglycin, n-formyl-glycine
PubChem CID 75606
IUPAC Name 2-formamidoacetic acid
SMILES OC(=O)CNC=O

Specifications

Melting Point 147°C to 151°C
Linear Formula HCONHCH2CO2H
Quantity 1 g
Formula Weight 103.08
Percent Purity 98%
Chemical Name or Material N-Formylglycine
EINECSNumber 219-649-8
RTECSNumber MC0547000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only