missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-tyrosine 98.0+%, TCI America™

Catalog No. p-7129505
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
F01351G 1 g
F01355G 5 g
2 options

Catalog No. F01351G

Supplier: TCI America F01351G

Only null left

Chemical Identifiers

CAS 13200-86-7
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.20
MDL Number MFCD00021031
InChI Key ROUWPHMRHBMAFE-UHFFFAOYNA-N
PubChem CID 13025455
ChEBI CHEBI:50603
IUPAC Name 3-(4-hydroxyphenyl)-2-formamidopropanoic acid
SMILES OC(=O)C(CC1=CC=C(O)C=C1)NC=O

Specifications

Melting Point 165°C
Color White
Quantity 1 g
Formula Weight 209.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Formyl-L-tyrosine