accessibility menu, dialog, popup

UserName

CAS RN 13200-86-7

OHOHOHNO

CAS RN 13200-86-7

IUPAC Name: 3-(4-hydroxyphenyl)-2-formamidopropanoic acid
Synonyms: 3-(4-hydroxyphenyl)-2-formamidopropanoic acid
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: ROUWPHMRHBMAFE-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=C(O)C=C1)NC=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results