missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-tyrosine 98.0+%, TCI America™

Catalog No. F01355G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01355G 5 g
F01351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F01355G Supplier TCI America Supplier No. F01355G

Chemical Identifiers

CAS 13200-86-7
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.20
MDL Number MFCD00021031
InChI Key ROUWPHMRHBMAFE-UHFFFAOYNA-N
PubChem CID 13025455
ChEBI CHEBI:50603
IUPAC Name 3-(4-hydroxyphenyl)-2-formamidopropanoic acid
SMILES OC(=O)C(CC1=CC=C(O)C=C1)NC=O

Specifications

Melting Point 165°C
Color White
Quantity 5 g
Formula Weight 209.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Formyl-L-tyrosine