missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-methionine 95.0+%, TCI America™

Catalog No. F01241G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F01241G Supplier TCI America Supplier No. F01241G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4289-98-9
Molecular Formula C6H11NO3S
Molecular Weight (g/mol) 177.22
MDL Number MFCD00021033
InChI Key PYUSHNKNPOHWEZ-YFKPBYRVSA-N
Synonym n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl methionine
PubChem CID 439750
ChEBI CHEBI:16552
IUPAC Name (2S)-2-formamido-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@H](NC=O)C(O)=O

Specifications

Melting Point 101°C
Color White
Quantity 1 g
Formula Weight 177.22
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Formyl-L-methionine