Learn More
N-Formyl-L-leucine, tech. 90%
CAS: 6113-61-7 | C7H13NO3 | 159.19 g/mol
$34.59 - $115.99
Chemical Identifiers
| CAS | 6113-61-7 |
|---|---|
| Molecular Formula | C7H13NO3 |
| Molecular Weight (g/mol) | 159.19 |
| MDL Number | MFCD00055861 |
| InChI Key | HFBHOAHFRNLZGN-MDOHGIEYNA-N |
| Synonym | n-formyl-l-leucine, n-formyl-leu-oh, n-formylleucine, l-leucine, n-formyl, unii-5tha63y4ff, 5tha63y4ff, 2s-2-formamido-4-methylpentanoic acid, n-formylleucine #, formylleucin, n-formyl leucine |
| PubChem CID | 9880216 |
| IUPAC Name | (2S)-2-formamido-4-methylpentanoic acid |
| SMILES | CC(C)C[C@H](NC=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3356403
|
Thermo Scientific Chemicals
H3356403 |
1 g |
Each for $34.59
|
|
|||||
|
AAH3356406
|
Thermo Scientific Chemicals
H3356406 |
5 g |
Each for $115.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6113-61-7 | |
| 159.19 | |
| HFBHOAHFRNLZGN-MDOHGIEYNA-N | |
| 9880216 | |
| CC(C)C[C@H](NC=O)C(O)=O |
| C7H13NO3 | |
| MFCD00055861 | |
| n-formyl-l-leucine, n-formyl-leu-oh, n-formylleucine, l-leucine, n-formyl, unii-5tha63y4ff, 5tha63y4ff, 2s-2-formamido-4-methylpentanoic acid, n-formylleucine #, formylleucin, n-formyl leucine | |
| (2S)-2-formamido-4-methylpentanoic acid |
Specifications
| 142°C to 145°C | |
| C7H13NO3 | |
| n-formyl-l-leucine, n-formyl-leu-oh, n-formylleucine, l-leucine, n-formyl, unii-5tha63y4ff, 5tha63y4ff, 2s-2-formamido-4-methylpentanoic acid, n-formylleucine #, formylleucin, n-formyl leucine | |
| CC(C)C[C@H](NC=O)C(O)=O | |
| 1 g | |
| 9880216 | |
| 90% | |
| N-Formyl-L-leucine |
| 6113-61-7 | |
| MFCD00055861 | |
| HFBHOAHFRNLZGN-MDOHGIEYNA-N | |
| (2S)-2-formamido-4-methylpentanoic acid | |
| 159.19 | |
| 159.18 | |
| Technical |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-080-4
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only