accessibility menu, dialog, popup

UserName

H3CCH3(S)OHHNOO

CAS RN 6113-61-7

IUPAC Name: (2S)-2-formamido-4-methylpentanoic acid
Synonyms: N-formylleucine
Molecular Weight (g/mol): 159.19
Molecular Formula: C7H13NO3
InChi Key: HFBHOAHFRNLZGN-MDOHGIEYNA-N
SMILES: CC(C)C[C@H](NC=O)C(O)=O
Quantity 
  • (2)
  • (1)
Grade 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results