missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-leucine, tech. 90%

Catalog No. AAH3356406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3356406 5 g
AAH3356403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3356406 Supplier Thermo Scientific Chemicals Supplier No. H3356406
Only null left

CAS: 6113-61-7 | C7H13NO3 | 159.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6113-61-7
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00055861
InChI Key HFBHOAHFRNLZGN-MDOHGIEYNA-N
Synonym n-formyl-l-leucine, n-formyl-leu-oh, n-formylleucine, l-leucine, n-formyl, unii-5tha63y4ff, 5tha63y4ff, 2s-2-formamido-4-methylpentanoic acid, n-formylleucine #, formylleucin, n-formyl leucine
PubChem CID 9880216
IUPAC Name (2S)-2-formamido-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC=O)C(O)=O

Specifications

Melting Point 142°C to 145°C
Quantity 5 g
Formula Weight 159.18
Percent Purity 90%
Grade Technical
Chemical Name or Material N-Formyl-L-leucine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-080-4
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only