missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-alanine 98.0+%, TCI America™

Catalog No. F01081G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F01081G Supplier TCI America Supplier No. F01081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10512-86-4
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.104
MDL Number MFCD00037357
InChI Key SRBATDDRZARFDZ-VKHMYHEASA-N
PubChem CID 1810758
IUPAC Name (2S)-2-formamidopropanoic acid
SMILES CC(C(=O)O)NC=O

Specifications

Color White
Quantity 1 g
Formula Weight 117.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Formyl-L-alanine