accessibility menu, dialog, popup

UserName

CH3(S)NHOOHO

CAS RN 10512-86-4

IUPAC Name: (2S)-2-formamidopropanoic acid
Synonyms: (2S)-2-formamidopropanoic acid
Molecular Weight (g/mol): 117.1
Molecular Formula: C4H7NO3
InChi Key: SRBATDDRZARFDZ-VKHMYHEASA-N
SMILES: C[C@H](NC=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results