missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-DL-2-aminobutyric Acid 98.0+%, TCI America™

Catalog No. F01091G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F01091G Supplier TCI America Supplier No. F01091G
Only null left

Chemical Identifiers

CAS 106873-99-8
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00059813
InChI Key QAISXTKWTBOYAA-UHFFFAOYNA-N
PubChem CID 559455
IUPAC Name 2-formamidobutanoic acid
SMILES CCC(NC=O)C(O)=O

Specifications

Melting Point 155°C
Color White
Quantity 1 g
Formula Weight 131.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Formyl-DL-2-aminobutyric Acid