accessibility menu, dialog, popup

UserName

CAS RN 106873-99-8

H3COHOHNO

CAS RN 106873-99-8

IUPAC Name: 2-formamidobutanoic acid
Synonyms: 2-formamidobutanoic acid
Molecular Weight (g/mol): 131.13
Molecular Formula: C5H9NO3
InChi Key: QAISXTKWTBOYAA-UHFFFAOYNA-N
SMILES: CCC(NC=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results